accessibility menu, dialog, popup

UserName

H3CNH3CCH2

CAS RN 5666-17-1

IUPAC Name: diethyl(prop-2-en-1-yl)amine
Synonyms: diethyl(prop-2-en-1-yl)amine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: JWAJUTZQGZBKFS-UHFFFAOYSA-N
SMILES: CCN(CC)CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results