CAS RN 566-24-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isochenodeoxycholic Acid, TRC
CAS: 566-24-5 Molecular Formula: C24H40O4 Molecular Weight (g/mol): 392.57 Synonym: (3β,5β,7α)-3,7-Dihydroxycholan-24-oic Acid,3β,7α-Dihydroxy-5β-cholan-24-oic Acid,3β,7α-Dihydroxy-5β-cholanic Acid,3β,7α-Dihydroxy-5β-cholanoic Acid,3β-Chenodeoxycholic Acid,BAR601,Isochenodeoxycholic Acid; SMILES: O[C@H]1CC[C@@]2(C)[C@@](C[C@@H](O)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCC(O)=O)([H])C1