accessibility menu, dialog, popup

UserName

CH3OOCH3Cl

CAS RN 56525-63-4

IUPAC Name: methyl 3-chloro-4-methylbenzoate
Synonyms: methyl 3-chloro-4-methylbenzoate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: KTFQDZCNPGFKAH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=C(C)C=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)