CAS RN 56525-09-8
CAS RN 56525-09-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitrosoatrazine, TRC
CAS: 56525-09-8 Molecular Formula: C8 H13 Cl N6 O Molecular Weight (g/mol): 244.68 Synonym: 6-Chloro-N2-ethyl-N4-(1-methylethyl)-N2-nitroso-1,3,5-triazine-2,4-diamine,6-Chloro-N-ethyl-N'-(1-methylethyl)-N-nitroso-1,3,5-triazine-2,4-diamine; IUPAC Name: N-[4-chloro-6-(propan-2-ylamino)-1,3,5-triazin-2-yl]-N-ethylnitrous amide SMILES: CCN(N=O)c1nc(Cl)nc(NC(C)C)n1