accessibility menu, dialog, popup

UserName

CH3NNOH3CCH3

CAS RN 56430-08-1

IUPAC Name: 1,5-dimethyl-2-(4-methylphenyl)-2,3-dihydro-1H-pyrazol-3-one
Synonyms: 1,5-dimethyl-2-(4-methylphenyl)pyrazol-3-one
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: CSVSLJZHBKTVNT-UHFFFAOYSA-N
SMILES: CN1N(C(=O)C=C1C)C1=CC=C(C)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results