accessibility menu, dialog, popup

UserName

CAS RN 563-80-4

H3CCH3CH3O

CAS RN 563-80-4

IUPAC Name: 3-methylbutan-2-one
Synonyms: methyl isopropyl ketone
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: SYBYTAAJFKOIEJ-UHFFFAOYSA-N
SMILES: CC(C)C(C)=O
Boiling Point 
  • (4)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)