CAS RN 563-71-3
CAS RN 563-71-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ferronil, TRC
CAS: 563-71-3 Molecular Formula: FeCO3 Molecular Weight (g/mol): 115.85 Synonym: Ferrous Carbonate,Ferrous Monocarbonate,Iron Carbonate (FeCO3),Iron(2+) Carbonate,Iron(II) Carbonate; IUPAC Name: iron(2+);carbonate SMILES: [Fe+2].[O-]C(=O)[O-]