CAS RN 5619-09-0
IUPAC Name:
methyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Synonyms:
methyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Molecular Weight (g/mol):
254.71
Molecular Formula:
C12H15ClN2O2
InChi Key:
XNFNGGQRDXFYMM-UHFFFAOYNA-N
SMILES:
Cl.COC(=O)C(N)CC1=CNC2=CC=CC=C12
1
–
1
of
1
results