CAS RN 5619-05-6
IUPAC Name:
methyl 2-amino-3-methylbutanoate hydrochloridyl
Synonyms:
methyl 2-amino-3-methylbutanoate hydrochloridyl
Molecular Weight (g/mol):
166.63
Molecular Formula:
C6H13ClNO2
InChi Key:
RFMIEHOYULVMNZ-UHFFFAOYNA-N
SMILES:
[Cl].COC(=O)C(N)C(C)C
1
–
1
of
1
results
1
DL-Valine methyl ester hydrochloride, 99%
CAS: 5619-05-6 MDL Number: MFCD00066118