accessibility menu, dialog, popup

UserName

CAS RN 56182-43-5

SNBr

CAS RN 56182-43-5

IUPAC Name: 2-bromothiophene-3-carbonitrile
Synonyms: 2-bromothiophene-3-carbonitrile
Molecular Weight (g/mol): 188.04
Molecular Formula: C5H2BrNS
InChi Key: GHMDOVNPFWGFSZ-UHFFFAOYSA-N
SMILES: BrC1=C(C=CS1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results