accessibility menu, dialog, popup

UserName

CAS RN 5616-51-3

CH3SS

CAS RN 5616-51-3

IUPAC Name: 2-methyl-1,3-dithiolane
Synonyms: 1,3-dithiolane, 2-methyl-
Molecular Weight (g/mol): 120.23
Molecular Formula: C4H8S2
InChi Key: CARJCVDELAMAEJ-UHFFFAOYSA-N
SMILES: CC1SCCS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results