accessibility menu, dialog, popup

UserName

CAS RN 56131-49-8

IUPAC Name: 4-cyanophenyl 4-propylbenzoate
Synonyms: 4-cyanophenyl 4-propylbenzoate
Molecular Weight (g/mol): 265.31
Molecular Formula: C17H15NO2
InChi Key: NCTWNVKZQMMLIV-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C(=O)OC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results