accessibility menu, dialog, popup

UserName

CAS RN 5612-67-9

CH3OO

CAS RN 5612-67-9

IUPAC Name: 2-methylfuran-3-carbaldehyde
Synonyms: 2-methylfuran-3-carbaldehyde
Molecular Weight (g/mol): 110.11
Molecular Formula: C6H6O2
InChi Key: WBFUBNWKSDINIX-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results