accessibility menu, dialog, popup

UserName

N(R)NPNHN(R)OPOOP(R)(S)OOOOOOOH2NOHHOOHHONaNa

CAS RN 56001-37-7

IUPAC Name: ({[(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(phosphonatooxy)phosphinate
Synonyms: {[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy(phosphonatooxy)phosphinate
Molecular Weight (g/mol): 519.15
Molecular Formula: C10H12N5O14P3
InChi Key: XKMLYUALXHKNFT-UUOKFMHZSA-J
SMILES: NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (9)
  • (4)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (9)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)