accessibility menu, dialog, popup

UserName

CAS RN 56-89-3

NH2(R)SS(R)NH2HOOOHO

CAS RN 56-89-3

IUPAC Name: (2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
Synonyms: L-cystine β,β'-dithioalanine, L- b,b'-dithiodialanine gelucystine L-dicysteine 1-cystine L cystine oxidized L-cysteine copper cystinate 3,3'-dithiobis
Molecular Weight (g/mol): 240.29
Molecular Formula: C6H12N2O4S2
InChi Key: LEVWYRKDKASIDU-IMJSIDKUSA-N
SMILES: N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (22)
  • (3)
Quantity 
  • (1)
  • (6)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
Grade 
  • (4)
  • (4)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (4)
  • (5)
  • (7)
  • (4)
  • (2)
  • (1)
  • (1)
  • (22)
  • (3)
  • (1)
  • (6)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)