accessibility menu, dialog, popup

UserName

CAS RN 56-57-5

ONONO

CAS RN 56-57-5

IUPAC Name: 4-nitroquinolin-1-ium-1-olate
Synonyms: 4-nitroquinoline-1-oxide
Molecular Weight (g/mol): 190.16
Molecular Formula: C9H6N2O3
InChi Key: YHQDZJICGQWFHK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C=CC=CC2=[N+]([O-])C=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)