CAS RN 56-41-7
L-Alanine, 99%
CAS: 56-41-7 Molecular Formula: C3H7NO2 Molecular Weight (g/mol): 89.09 MDL Number: MFCD00064410 InChI Key: QNAYBMKLOCPYGJ-REOHCLBHSA-N Synonym: l-alanine,alanine,l-alpha-alanine,s-alanine,s-2-aminopropanoic acid,l-2-aminopropionic acid,2s-2-aminopropanoic acid,h-ala-oh,l-+-alanine,l-2-aminopropanoic acid PubChem CID: 5950 ChEBI: CHEBI:16977 IUPAC Name: (2S)-2-aminopropanoic acid SMILES: C[C@H](N)C(O)=O
Thermo Scientific Chemicals L-Alanine, 99%
CAS: 56-41-7 Molecular Formula: C3H7NO2 Molecular Weight (g/mol): 89.09 MDL Number: MFCD00064410 InChI Key: QNAYBMKLOCPYGJ-REOHCLBHSA-N Synonym: l-alanine,alanine,l-alpha-alanine,s-alanine,s-2-aminopropanoic acid,l-2-aminopropionic acid,2s-2-aminopropanoic acid,h-ala-oh,l-+-alanine,l-2-aminopropanoic acid PubChem CID: 5950 ChEBI: CHEBI:16977 IUPAC Name: (2S)-2-aminopropanoic acid SMILES: C[C@H](N)C(O)=O
Thermo Scientific Chemicals L-Alanine, Cell Culture Reagent
CAS: 56-41-7 Molecular Formula: C3H7NO2 Molecular Weight (g/mol): 89.09 MDL Number: MFCD00064410 InChI Key: QNAYBMKLOCPYGJ-REOHCLBHSA-N Synonym: l-alanine,alanine,l-alpha-alanine,s-alanine,s-2-aminopropanoic acid,l-2-aminopropionic acid,2s-2-aminopropanoic acid,h-ala-oh,l-+-alanine,l-2-aminopropanoic acid PubChem CID: 5950 ChEBI: CHEBI:16977 IUPAC Name: (2S)-2-aminopropanoic acid SMILES: C[C@H](N)C(O)=O
L-Alanine, 99%, specified according to the requirements of Ph.Eur.
CAS: 56-41-7 Molecular Formula: C3H7NO2 Molecular Weight (g/mol): 89.09 MDL Number: MFCD00064410 InChI Key: QNAYBMKLOCPYGJ-REOHCLBHSA-N Synonym: l-alanine,alanine,l-alpha-alanine,s-alanine,s-2-aminopropanoic acid,l-2-aminopropionic acid,2s-2-aminopropanoic acid,h-ala-oh,l-+-alanine,l-2-aminopropanoic acid PubChem CID: 5950 ChEBI: CHEBI:16977 IUPAC Name: (2S)-2-aminopropanoic acid SMILES: C[C@H](N)C(O)=O
L-Alanine, 99%, MP Biomedicals™
CAS: 56-41-7 Molecular Formula: C3H7NO2 Molecular Weight (g/mol): 89.09 MDL Number: MFCD00064410 InChI Key: QNAYBMKLOCPYGJ-REOHCLBHSA-N Synonym: l-alanine,alanine,l-alpha-alanine,s-alanine,s-2-aminopropanoic acid,l-2-aminopropionic acid,2s-2-aminopropanoic acid,h-ala-oh,l-+-alanine,l-2-aminopropanoic acid PubChem CID: 5950 ChEBI: CHEBI:16977 IUPAC Name: (2S)-2-aminopropanoic acid SMILES: C[C@H](N)C(O)=O
L-Alanine, MP Biomedicals™
CAS: 56-41-7 Molecular Formula: C3H7NO2 Molecular Weight (g/mol): 89.09 MDL Number: MFCD00064410 InChI Key: QNAYBMKLOCPYGJ-REOHCLBHSA-N Synonym: l-alanine,alanine,l-alpha-alanine,s-alanine,s-2-aminopropanoic acid,l-2-aminopropionic acid,2s-2-aminopropanoic acid,h-ala-oh,l-+-alanine,l-2-aminopropanoic acid PubChem CID: 5950 ChEBI: CHEBI:16977 IUPAC Name: (2S)-2-aminopropanoic acid SMILES: C[C@H](N)C(O)=O
Melford L-Alanine
L-Alanine is a crucial building block for protein synthesis.
(S)-(+)-Alanine, Sigma-Aldrich™
CAS: 56-41-7 Grade: Research Molecular Weight (g/mol): 89.09
Alanine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
l-Alanine, Millipore™
CAS: 56-41-7
L-Alanine, TRC
CAS: 56-41-7 Molecular Formula: C3 H7 N O2 Molecular Weight (g/mol): 89.09 Synonym: Alanine, L- (7CI,8CI),(2S)-2-Aminopropanoic acid,(S)-(+)-Alanine,(S)-2-Aminopropanoic acid,(S)-Alanine,2-Aminopropionic acid,Alanine,L-(+)-Alanine,L-2-Aminopropanoic acid,L-2-Aminopropionic acid,L-α-Alanine,L-α-Aminopropionic acid,Lactamine,NSC 206315,Propanoic acid, 2-amino-, (S)-,α-Alanine,α-Aminopropionic acid IUPAC Name: (2S)-2-aminopropanoic acid SMILES: C[C@H](N)C(=O)O
L-Alanine, MedChemExpress
MedChemExpress L-Alanine is a non-essential amino acid, involved in sugar and acid metabolism, increases immunity, and provides energy for muscle tissue, brain, and central nervous system.
Alanine (L-Alanine), LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
- 1
- 2