CAS RN 55837-27-9
CAS RN 55837-27-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Piretanide, TRC
CAS: 55837-27-9 Molecular Formula: C17 H18 N2 O5 S Molecular Weight (g/mol): 362.4 Synonym: Piretanide,4-Phenoxy-3-(pyrrolidin-1-yl)-5-sulfamoylbenzoic acid IUPAC Name: 4-phenoxy-3-pyrrolidin-1-yl-5-sulfamoylbenzoic acid SMILES: NS(=O)(=O)c1cc(cc(N2CCCC2)c1Oc3ccccc3)C(=O)O