accessibility menu, dialog, popup

UserName

CAS RN 558-32-7

IUPAC Name: hexafluoro-λ⁵-phosphanuide; tetramethylazanium
Synonyms: tetramethylammonium ion hexafluorophosphate
Molecular Weight (g/mol): 219.11
Molecular Formula: C4H12F6NP
InChi Key: ZFKULDQWLXAKIM-UHFFFAOYSA-N
SMILES: C[N+](C)(C)C.F[P-](F)(F)(F)(F)F
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results