accessibility menu, dialog, popup

UserName

CAS RN 558-13-4

BrBrBrBr

CAS RN 558-13-4

IUPAC Name: tetrabromomethane
Synonyms: carbon tetrabromide kohlenstofftetrabromid carbon bromide CBr4 tetrabromkohlenstoff
Molecular Weight (g/mol): 331.63
Molecular Formula: CBr4
InChi Key: HJUGFYREWKUQJT-UHFFFAOYSA-N
SMILES: BrC(Br)(Br)Br
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (3)