accessibility menu, dialog, popup

UserName

HONNHNOOH

CAS RN 55779-48-1

IUPAC Name: 8-benzyl-6-(4-hydroxyphenyl)-2-[(4-hydroxyphenyl)methyl]-3H,7H-imidazo[1,2-a]pyrazin-3-one
Synonyms: coelenterazine coelenterate luciferin oplophorus luciferin
Molecular Weight (g/mol): 423.47
Molecular Formula: C26H21N3O3
InChi Key: YHIPILPTUVMWQT-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC2=NC3=C(CC4=CC=CC=C4)NC(=CN3C2=O)C2=CC=C(O)C=C2)C=C1
Quantity 
  • (3)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results