accessibility menu, dialog, popup

UserName

CAS RN 557-91-5

CH3BrBr

CAS RN 557-91-5

IUPAC Name: 1,1-dibromoethane
Synonyms: dibromide, ethylene
Molecular Weight (g/mol): 187.86
Molecular Formula: C2H4Br2
InChi Key: APQIUTYORBAGEZ-UHFFFAOYSA-N
SMILES: CC(Br)Br
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)