CAS RN 557-21-1
CAS RN 557-21-1
IUPAC Name:
zinc cyanide
Synonyms:
zinc cyanide
Molecular Weight (g/mol):
117.42
Molecular Formula:
C2N2Zn
InChi Key:
GTLDTDOJJJZVBW-UHFFFAOYSA-N
SMILES:
[Zn++].[C-]#N.[C-]#N
1
–
3
of
3
results
1
Zinc cyanide, 98%
CAS: 557-21-1 Molecular Formula: Zn(CN)2 MDL Number: MFCD00011292
2
Zinc cyanide, 98%, pure
CAS: 557-21-1 Molecular Formula: C2N2Zn Molecular Weight (g/mol): 117.43 MDL Number: MFCD00011292
3
Zinc cyanide, 85%
CAS: 557-21-1 MDL Number: MFCD00011292