accessibility menu, dialog, popup

UserName

MgH3COOH3COO

CAS RN 557-04-0

IUPAC Name: magnesium(2+) dioctadecanoate
Synonyms: magnesium stearate magnesium distearate magnesium octadecanoate
Molecular Weight (g/mol): 591.26
Molecular Formula: C36H70MgO4
InChi Key: HQKMJHAJHXVSDF-UHFFFAOYSA-L
SMILES: [Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)