CAS RN 556801-15-1
CAS RN 556801-15-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Desfluorolinezolid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Defluoro Linezolid, TRC
CAS: 556801-15-1 Molecular Formula: C16H21N3O4 Molecular Weight (g/mol): 319.36 Synonym: Acetamide, N-[[(5S)-3-[4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]-,Desfluorolinezolid IUPAC Name: N-[[(5S)-3-(4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide SMILES: CC(=O)NC[C@H]1CN(C(=O)O1)c2ccc(cc2)N3CCOCC3