accessibility menu, dialog, popup

UserName

CAS RN 556-24-1

H3COOCH3CH3

CAS RN 556-24-1

IUPAC Name: methyl 3-methylbutanoate
Synonyms: methyl isovalerate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: OQAGVSWESNCJJT-UHFFFAOYSA-N
SMILES: COC(=O)CC(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results