accessibility menu, dialog, popup

UserName

CAS RN 55563-79-6

IUPAC Name: pent-4-en-2-ol
Synonyms: 4-penten-2-ol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: ZHZCYWWNFQUZOR-UHFFFAOYNA-N
SMILES: CC(O)CC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results