CAS RN 555-57-7
CAS RN 555-57-7
IUPAC Name:
benzyl(methyl)(prop-2-yn-1-yl)amine
Synonyms:
pargyline pargyline preparation pargyline (substance) pargylin paragyline pargyline (product) pargylinum pargilina benzyl-methyl-2-propinylamin methylbenzylpropynylamine
Molecular Weight (g/mol):
159.23
Molecular Formula:
C11H13N
InChi Key:
DPWPWRLQFGFJFI-UHFFFAOYSA-N
SMILES:
CN(CC#C)CC1=CC=CC=C1
No Refinements
1
–
0
of
0
results
1
N-Methyl-N-propargylbenzylamine, 97%
CAS: 555-57-7 Molecular Formula: C11H13N Molecular Weight (g/mol): 159.23 MDL Number: MFCD00008576 InChI Key: DPWPWRLQFGFJFI-UHFFFAOYSA-N Synonym: pargyline,pargylamine,n-methyl-n-propargylbenzylamine,paragyline,eudatin,supirdyl,eutonyl,pargylin,n-methyl-n-2-propynylbenzylamine,benzyl-methyl-2-propinylamin PubChem CID: 4688 ChEBI: CHEBI:7930 IUPAC Name: N-benzyl-N-methylprop-2-yn-1-amine SMILES: CN(CC#C)CC1=CC=CC=C1