accessibility menu, dialog, popup

UserName

CAS RN 555-06-6

NaNH2OO

CAS RN 555-06-6

IUPAC Name: sodium 4-aminobenzoate
Synonyms: sodium p-aminobenzoate
Molecular Weight (g/mol): 159.12
Molecular Formula: C7H6NNaO2
InChi Key: XETSAYZRDCRPJY-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC=C(C=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results