accessibility menu, dialog, popup

UserName

CAS RN 55357-38-5

CH3SOOOCH3NCH3CH3HO

CAS RN 55357-38-5

IUPAC Name: (2-hydroxyethyl)trimethylazanium 4-methylbenzene-1-sulfonate
Synonyms: choline tosylate
Molecular Weight (g/mol): 275.36
Molecular Formula: C12H21NO4S
InChi Key: DVGVMQVOCJNXNJ-UHFFFAOYSA-M
SMILES: C[N+](C)(C)CCO.CC1=CC=C(C=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results