accessibility menu, dialog, popup

UserName

CAS RN 553-90-2

IUPAC Name: dimethyl oxalate
Synonyms: dimethyl oxalate
Molecular Weight (g/mol): 118.09
Molecular Formula: C4H6O4
InChi Key: LOMVENUNSWAXEN-UHFFFAOYSA-N
SMILES: COC(=O)C(=O)OC
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)