accessibility menu, dialog, popup

UserName

CAS RN 553-86-6

OO

CAS RN 553-86-6

IUPAC Name: 2,3-dihydro-1-benzofuran-2-one
Synonyms: 3H-benzofuran-2-one
Molecular Weight (g/mol): 134.13
Molecular Formula: C8H6O2
InChi Key: ACZGCWSMSTYWDQ-UHFFFAOYSA-N
SMILES: O=C1CC2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)