accessibility menu, dialog, popup

UserName

H3COClCl

CAS RN 553-82-2

IUPAC Name: 2,4-dichloro-1-methoxybenzene
Synonyms: 2,4-dichloro-1-methoxybenzene
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: CICQUFBZCADHHX-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results