accessibility menu, dialog, popup

UserName

CH3OOSHNOCl

CAS RN 551910-46-4

IUPAC Name: methyl 2-(3-chloropropanamido)-4H,5H,6H-cyclopenta[b]thiophene-3-carboxylate
Synonyms: methyl 2-(3-chloropropanamido)-4H,5H,6H-cyclopenta[b]thiophene-3-carboxylate
Molecular Weight (g/mol): 287.76
Molecular Formula: C12H14ClNO3S
InChi Key: OMCRFVOYMSUXGQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(NC(=O)CCCl)SC2=C1CCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results