accessibility menu, dialog, popup

UserName

H3COClO

CAS RN 55150-29-3

IUPAC Name: 2-ethoxynaphthalene-1-carbonyl chloride
Synonyms: 2-ethoxynaphthalene-1-carbonyl chloride
Molecular Weight (g/mol): 234.68
Molecular Formula: C13H11ClO2
InChi Key: ZRVFFIXOCFUHDA-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2C=CC=CC2=C1C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results