accessibility menu, dialog, popup

UserName

CAS RN 55052-28-3

HNNCl

CAS RN 55052-28-3

IUPAC Name: 4-chloro-1H-pyrrolo[2,3-b]pyridine
Synonyms: 4-chloro-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: HNTZVGMWXCFCTA-UHFFFAOYSA-N
SMILES: ClC1=C2C=CNC2=NC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)