accessibility menu, dialog, popup

UserName

H3CSOOCl

CAS RN 54997-92-1

IUPAC Name: 4-butylbenzene-1-sulfonyl chloride
Synonyms: 4-butylbenzenesulfonyl chloride
Molecular Weight (g/mol): 232.72
Molecular Formula: C10H13ClO2S
InChi Key: OVFZELSNOHIDEF-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)