accessibility menu, dialog, popup

UserName

CAS RN 5495-84-1

IUPAC Name: 2-(propan-2-yl)-9H-thioxanthen-9-one
Synonyms: isopropyl thioxanthone
Molecular Weight (g/mol): 254.35
Molecular Formula: C16H14OS
InChi Key: KTALPKYXQZGAEG-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C2SC3=CC=CC=C3C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results