CAS RN 5470-95-1
CAS RN 5470-95-1
IUPAC Name:
(E)-N-[(2,3-dimethoxyphenyl)methylidene]hydroxylamine
Synonyms:
(E)-N-[(2,3-dimethoxyphenyl)methylidene]hydroxylamine
Molecular Weight (g/mol):
181.19
Molecular Formula:
C9H11NO3
InChi Key:
GKAIQNACVNFHCU-UXBLZVDNSA-N
SMILES:
COC1=CC=CC(\C=N\O)=C1OC
1
–
1
of
1
results
1
2,3-Dimethoxybenzaldoxime, 97%, Thermo Scientific™
CAS: 5470-95-1 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00033234 InChI Key: GKAIQNACVNFHCU-UXBLZVDNSA-N Synonym: 2,3-dimethoxybenzaldoxime,o-veratraldehyde, oxime,benzaldehyde, 2,3-dimethoxy-, oxime,e-n-2,3-dimethoxyphenyl methylidene hydroxylamine,ne-n-2,3-dimethoxyphenyl methylidene hydroxylamine,z-2,3-dimethoxybenzaldehyde oxime,benzaldehyde, 2,3-dimethoxy-,oxime,2-08-00-00269 beilstein handbook reference,1e-2,3-dimethoxybenzaldehyde oxime,2,3-dimethoxyphenyl hydroxyimino methane PubChem CID: 6871292 IUPAC Name: (NE)-N-[(2,3-dimethoxyphenyl)methylidene]hydroxylamine SMILES: COC1=CC=CC(\C=N\O)=C1OC