accessibility menu, dialog, popup

UserName

CAS RN 5469-66-9

IUPAC Name: 3-[(4-methylbenzenesulfonyl)oxy]propyl 4-methylbenzene-1-sulfonate
Synonyms: 3-[(4-methylbenzenesulfonyl)oxy]propyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 384.46
Molecular Formula: C17H20O6S2
InChi Key: ODVREDGIWSDQFD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCCOS(=O)(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results