accessibility menu, dialog, popup

UserName

CAS RN 5466-89-7

CH3H3COOHHO

CAS RN 5466-89-7

IUPAC Name: 1-(3,4-dihydroxyphenyl)-2-methylpropan-1-one
Synonyms: 1-(3,4-dihydroxyphenyl)-2-methylpropan-1-one
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: VDQLKIBLTMPAHI-UHFFFAOYSA-N
SMILES: CC(C)C(=O)C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results