accessibility menu, dialog, popup

UserName

CAS RN 54656-96-1

H3CNOH

CAS RN 54656-96-1

IUPAC Name: 1-(pyridin-4-yl)ethan-1-ol
Synonyms: 1-(pyridin-4-yl)ethanol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: HVOAMIOKNARIMR-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results