accessibility menu, dialog, popup

UserName

CAS RN 5462-82-8

IUPAC Name: phenanthrene-4,5-dicarboxylic acid
Synonyms: phenanthrene-4,5-dicarboxylic acid
Molecular Weight (g/mol): 266.25
Molecular Formula: C16H10O4
InChi Key: RAGXPRCIICFJIK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C(C=CC3=CC=CC(C(O)=O)=C23)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results