accessibility menu, dialog, popup

UserName

CAS RN 546-93-0

MgOOO

CAS RN 546-93-0

IUPAC Name: magnesium(2+) carbonate
Synonyms: magnesium carbonate magnesiumkarbonat MgCO3 magnesium carbonate anhydrous
Molecular Weight (g/mol): 84.31
Molecular Formula: CMgO3
InChi Key: ZLNQQNXFFQJAID-UHFFFAOYSA-L
SMILES: [Mg++].[O-]C([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results