accessibility menu, dialog, popup

UserName

CH3HO

CAS RN 5445-24-9

IUPAC Name: 1-propylcyclohexan-1-ol
Synonyms: 1-propylcyclohexan-1-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: PYLPYOPJKOJRNP-UHFFFAOYSA-N
SMILES: CCCC1(O)CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results