CAS RN 5439-11-2
CAS RN 5439-11-2
IUPAC Name:
2-bromo-N-(2-chlorophenyl)acetamide
Synonyms:
2-bromo-N-(2-chlorophenyl)acetamide
Molecular Weight (g/mol):
248.5
Molecular Formula:
C8H7BrClNO
InChi Key:
WIOJXHICOOQVAB-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC=C1NC(=O)CBr
1
–
1
of
1
results
1
N1-(2-Chlorophenyl)-2-bromoacetamide, 95%, Thermo Scientific™
CAS: 5439-11-2 Molecular Formula: C8H7BrClNO Molecular Weight (g/mol): 248.50 MDL Number: MFCD00173815 InChI Key: WIOJXHICOOQVAB-UHFFFAOYSA-N Synonym: 2-bromo-n-2-chlorophenyl acetamide,n1-2-chlorophenyl-2-bromoacetamide,n-bromoacetyl-2'-chloroacetaniline,wiojxhicooqvab-uhfffaoysa-n,2-bromo-n-2-chloro-phenyl-acetamide,acetamide, 2-bromo-n-2-chlorophenyl PubChem CID: 225525