accessibility menu, dialog, popup

UserName

H3COOIHO

CAS RN 5438-36-8

IUPAC Name: 4-hydroxy-3-iodo-5-methoxybenzaldehyde
Synonyms: 4-hydroxy-3-iodo-5-methoxybenzaldehyde
Molecular Weight (g/mol): 278.05
Molecular Formula: C8H7IO3
InChi Key: FBBCSYADXYILEH-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC(I)=C1O
Molecular Weight (g/mol) 
  • (5)
  • (6)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (5)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (6)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)