accessibility menu, dialog, popup

UserName

CAS RN 54370-00-2

H3COBrOHOCH3

CAS RN 54370-00-2

IUPAC Name: (2-bromo-4,5-dimethoxyphenyl)methanol
Synonyms: (2-bromo-4,5-dimethoxyphenyl)methanol
Molecular Weight (g/mol): 247.09
Molecular Formula: C9H11BrO3
InChi Key: SDZSRNYOXRHPHZ-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(CO)C=C1OC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)