accessibility menu, dialog, popup

UserName

CAS RN 5431-44-7

IUPAC Name: pyridine-2,6-dicarbaldehyde
Synonyms: pyridine-2,6-dicarbaldehyde
Molecular Weight (g/mol): 135.12
Molecular Formula: C7H5NO2
InChi Key: PMWXGSWIOOVHEQ-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(C=O)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results