CAS RN 5422-63-9
CAS RN 5422-63-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ethyl 4-Formamidobenzoate (N-Formylbenzocaine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-Formyl Benzocaine, TRC
CAS: 5422-63-9 Molecular Formula: C10 H11 N O3 Molecular Weight (g/mol): 193.199 Synonym: 4-(Formylamino)benzoic Acid Ethyl Ester,Benzoic acid, p-formamido-, ethyl ester (6CI),Ethyl 4-(formylamino)benzoate,Ethyl 4-formamidobenzoate,Ethyl p-formamidobenzoate,NSC 12009,Ethyl 4-Formamidobenzoate,N-Formylbenzocaine,N-Formyl benzocaine IUPAC Name: ethyl 4-formamidobenzoate SMILES: CCOC(=O)c1ccc(NC=O)cc1